Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elovl7
Entrez Gene ID
361895
Gene Name
ELOVL fatty acid elongase 7
Gene Aliases
-
Chromosome Location
Chr.2: 40373211 - 40386643 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191844.1 NP_001178773.1 - 259 146

View Full Product Details