Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd32
Entrez Gene ID
294601
Gene Name
ankyrin repeat domain 32
Gene Aliases
-
Chromosome Location
Chr.2: 4095126 - 4195819 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106400.1 NP_001099870.1 2 - 3 219 95

View Full Product Details