The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017491.1 | NP_001017491.1 | 4 - 5 | 644 | 132 |
XM_006249793.2 | XP_006249855.1 | 4 - 5 | 779 | 132 | |
GenBank mRNA | BC089837.1 | - | 4 - 5 | 644 | 132 |