Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mdm4
Entrez Gene ID
304798
Gene Name
MDM4, p53 regulator
Gene Aliases
-
Chromosome Location
Chr.13: 49786776 - 49828786 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012026.1 NP_001012026.1 8 - 9 789 126
XM_006249777.2 XP_006249839.1 8 - 9 850 126
GenBank mRNA BC083647.1 - 8 - 9 789 126

View Full Product Details