Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnni1
Entrez Gene ID
29388
Gene Name
troponin I type 1 (skeletal, slow)
Gene Aliases
-
Chromosome Location
Chr.13: 52624856 - 52637297 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006249862.2 XP_006249924.1 6 - 7 2119 59
XM_006249864.1 XP_006249926.1 6 - 7 698 59
XM_006249865.2 XP_006249927.1 9 - 10 741 59
XM_006249866.2 XP_006249928.1 9 - 10 672 59
XM_006249868.2 XP_006249930.1 7 - 8 707 59
XM_006249869.2 XP_006249931.1 7 - 8 631 59
GenBank mRNA FQ214889.1 - 9 - 10 719 59
FQ216822.1 - 9 - 10 692 59

View Full Product Details