The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031082.1 | NP_112344.1 | 8 - 9 | 956 | 92 |
XM_006254691.2 | XP_006254753.1 | 8 - 9 | 2196 | 92 | |
XM_008772177.1 | XP_008770399.1 | 9 - 10 | 958 | 92 | |
GenBank mRNA | L24116.1 | - | 8 - 9 | 956 | 92 |