Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc26a2
Entrez Gene ID
117267
Gene Name
solute carrier family 26 (anion exchanger), member 2
Gene Aliases
Dtdst
Chromosome Location
Chr.18: 56518999 - 56534539 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254787.2 XP_006254849.1 2 - 3 481 131

View Full Product Details