Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sncaip
Entrez Gene ID
307309
Gene Name
synuclein, alpha interacting protein
Gene Aliases
Syph1
Chromosome Location
Chr.18: 47739369 - 47877679 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107379.1 NP_001100849.1 4 - 5 1372 85
XM_006254719.2 XP_006254781.1 4 - 5 1358 85
XM_006254720.2 XP_006254782.1 4 - 5 1109 85
XM_006254721.2 XP_006254783.1 4 - 5 1203 85

View Full Product Details