Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spink10
Entrez Gene ID
100361300
Gene Name
serine peptidase inhibitor, Kazal type 10
Gene Aliases
-
Chromosome Location
Chr.18: 57968119 - 57981976 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002728560.3 XP_002728606.3 5 - 6 360 101

View Full Product Details