Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prrc1
Entrez Gene ID
291444
Gene Name
proline-rich coiled-coil 1
Gene Aliases
Prcc1
Chromosome Location
Chr.18: 52423845 - 52460854 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033887.1 NP_001029059.1 1 - 2 128 88
GenBank mRNA BC101903.1 - 1 - 2 128 88

View Full Product Details