Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lag3
Entrez Gene ID
297596
Gene Name
lymphocyte-activation gene 3
Gene Aliases
CD223
Chromosome Location
Chr.4: 157425644 - 157433695 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212513.2 NP_997678.2 7 - 8 1526 96
XM_006237348.2 XP_006237410.1 7 - 8 1751 96
XM_008763282.1 XP_008761504.1 8 - 9 1632 96
XM_008763283.1 XP_008761505.1 6 - 7 1276 96
GenBank mRNA AY230414.1 - 7 - 8 1416 96
BC091201.1 - 7 - 8 1526 96
DQ438937.1 - 7 - 8 1404 96

View Full Product Details