Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec4a1
Entrez Gene ID
362430
Gene Name
C-type lectin domain family 4, member A1
Gene Aliases
Dcir4
Chromosome Location
Chr.4: 155947794 - 155959909 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005890.2 NP_001005890.1 3 - 4 427 163
GenBank mRNA AY397672.1 - 3 - 4 328 163
BC098830.1 - 3 - 4 427 163

View Full Product Details