Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpb1
Entrez Gene ID
24271
Gene Name
carboxypeptidase B1 (tissue)
Gene Aliases
Cpb
Chromosome Location
Chr.2: 105059426 - 105089659 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012533.1 NP_036665.1 7 - 8 682 78

View Full Product Details