The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127652.1 | NP_001121124.1 | - | 196 | 71 |
GenBank mRNA | BC158762.1 | - | - | 196 | 71 |
FQ216242.1 | - | 2 - 2 | 234 | 71 | |
FQ216641.1 | - | 2 - 2 | 236 | 71 | |
FQ224910.1 | - | 2 - 2 | 234 | 71 | |
X59736.1 | - | 2 - 2 | 224 | 71 |