Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ckmt2
Entrez Gene ID
688698
Gene Name
creatine kinase, mitochondrial 2, sarcomeric
Gene Aliases
-
Chromosome Location
Chr.2: 21422680 - 21452797 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127652.1 NP_001121124.1 - 1057 66
GenBank mRNA BC158762.1 - - 1057 66
FQ215460.1 - 5 - 6 709 66
FQ216242.1 - 5 - 6 885 66
FQ216641.1 - 7 - 8 1098 66
FQ224910.1 - 7 - 8 1096 66
X59736.1 - 7 - 8 1085 66

View Full Product Details