Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spz1
Entrez Gene ID
499510
Gene Name
spermatogenic leucine zipper 1
Gene Aliases
-
Chromosome Location
Chr.2: 22145457 - 22147053 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024297.1 NP_001019468.1 1 - 1 1399 69
GenBank mRNA BC079262.1 - 1 - 1 1399 69

View Full Product Details