Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppap2a
Entrez Gene ID
64369
Gene Name
phosphatidic acid phosphatase type 2A
Gene Aliases
-
Chromosome Location
Chr.2: 44664085 - 44726820 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF503610.1 - 1 - 2 50 136

View Full Product Details