Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fst
Entrez Gene ID
24373
Gene Name
follistatin
Gene Aliases
FOL1, Fst-288, RATFOL1
Chromosome Location
Chr.2: 46537589 - 46544812 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012561.1 NP_036693.1 2 - 3 273 91
XM_006231953.2 XP_006232015.1 2 - 3 633 91
XM_006231954.2 XP_006232016.1 2 - 3 633 91

View Full Product Details