Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tm4sf4
Entrez Gene ID
116467
Gene Name
transmembrane 4 L six family member 4
Gene Aliases
Lrtm4
Chromosome Location
Chr.2: 147532032 - 147547254 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053785.1 NP_446237.1 4 - 5 926 121
GenBank mRNA AF205717.1 - 4 - 5 926 121
BC091131.1 - 4 - 5 741 121

View Full Product Details