The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031526.1 | NP_113714.1 | 2 - 3 | 436 | 86 |
GenBank mRNA | X14211.1 | - | 2 - 3 | 341 | 86 |