Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fxr2
Entrez Gene ID
287433
Gene Name
fragile X mental retardation, autosomal homolog 2
Gene Aliases
Fxr2h
Chromosome Location
Chr.10: 56237814 - 56257877 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100647.1 NP_001094117.1 12 - 13 1355 152
GenBank mRNA BC160929.1 - 12 - 13 1355 152

View Full Product Details