Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cog1
Entrez Gene ID
303652
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
-
Chromosome Location
Chr.10: 102167502 - 102180569 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107062.2 NP_001100532.2 1 - 2 320 112
XM_006247686.2 XP_006247748.1 1 - 2 589 112
XM_006247687.2 XP_006247749.1 1 - 2 589 112
GenBank mRNA BC169068.1 - - 318 112
FQ212043.1 - 1 - 2 594 112

View Full Product Details