Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrpl12
Entrez Gene ID
303746
Gene Name
mitochondrial ribosomal protein L12
Gene Aliases
Rpm12
Chromosome Location
Chr.10: 109658032 - 109662535 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029900.1 NP_001025071.1 3 - 4 583 102
GenBank mRNA FQ228179.1 - 3 - 4 397 102
FQ233456.1 - 3 - 4 399 102
FQ234892.1 - 3 - 4 397 102

View Full Product Details