The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029900.1 | NP_001025071.1 | 4 - 5 | 712 | 111 |
GenBank mRNA | FQ228179.1 | - | 4 - 5 | 526 | 111 |
FQ233456.1 | - | 4 - 5 | 528 | 111 | |
FQ234892.1 | - | 4 - 5 | 526 | 111 |