Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galnt10
Entrez Gene ID
170501
Gene Name
polypeptide N-acetylgalactosaminyltransferase 10
Gene Aliases
Galnt9
Chromosome Location
Chr.10: 43067316 - 43208992 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130742.1 NP_570098.1 7 - 8 1050 129
GenBank mRNA AF241241.1 - 7 - 8 1050 129

View Full Product Details