Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nlk
Entrez Gene ID
497961
Gene Name
nemo like kinase
Gene Aliases
RGD1561602
Chromosome Location
Chr.10: 65840052 - 65963932 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191924.1 NP_001178853.1 2 - 3 1785 76
XM_006246945.2 XP_006247007.1 2 - 3 1587 76
GenBank mRNA FQ225006.1 - 2 - 3 640 76

View Full Product Details