Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnai2
Entrez Gene ID
360654
Gene Name
dynein, axonemal, intermediate chain 2
Gene Aliases
Dnaic2
Chromosome Location
Chr.10: 103266343 - 103301521 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007726.1 NP_001007727.1 2 - 3 547 65
GenBank mRNA BC081920.1 - 2 - 3 547 65

View Full Product Details