Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nthl1
Entrez Gene ID
29541
Gene Name
nth (endonuclease III)-like 1 (E.coli)
Gene Aliases
Nth1
Chromosome Location
Chr.10: 13996660 - 14002827 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105728.1 NP_001099198.1 3 - 4 573 89

View Full Product Details