The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144991.1 | NP_001138463.1 | 12 - 13 | 1959 | 69 |
XM_006246562.2 | XP_006246624.2 | 12 - 13 | 1977 | 69 | |
XM_006246563.2 | XP_006246625.2 | 12 - 13 | 1832 | 69 |