Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tvp23b
- Entrez Gene ID
-
687358
- Gene Name
- trans-golgi network vesicle protein 23 homolog B (S. cerevisiae)
- Gene Aliases
-
Fam18b2
- Chromosome Location
-
Chr.10: 49299125 - 49310402 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FQ212667.1 |
- |
1 - 2 |
81 |
96 |
View Full Product Details