Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Armcx1
Entrez Gene ID
501619
Gene Name
armadillo repeat containing, X-linked 1
Gene Aliases
-
Chromosome Location
Chr.X: 105911925 - 105915823 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024367.1 NP_001019538.1 - 40 54
GenBank mRNA BC085780.1 - - 40 54
FQ221160.1 - - 64 54

Gene Symbol
LOC102554790
Entrez Gene ID
102554790
Gene Name
armadillo repeat-containing X-linked protein 1-like
Gene Aliases
-
Chromosome Location
Chr.X: 105535587 - 105539575 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257242.2 XP_006257304.1 1 - 2 123 54

View Full Product Details