Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdx4
Entrez Gene ID
302400
Gene Name
caudal type homeo box 4
Gene Aliases
RGD1561529
Chromosome Location
Chr.X: 74165172 - 74173758 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106942.1 NP_001100412.1 1 - 2 683 127

View Full Product Details