Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cylc1
Entrez Gene ID
100359483
Gene Name
cylicin, basic protein of sperm head cytoskeleton 1
Gene Aliases
-
Chromosome Location
Chr.X: 82217133 - 82302106 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229306.2 XP_006229368.1 4 - 5 280 127
XM_006229307.2 XP_006229369.1 4 - 5 280 127

View Full Product Details