The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107187.1 | NP_001100657.1 | 5 - 6 | 950 | 91 |
XM_006250058.2 | XP_006250120.1 | 6 - 7 | 1069 | 91 | |
XM_006250059.2 | XP_006250121.1 | 7 - 8 | 1167 | 91 | |
XM_008769634.1 | XP_008767856.1 | 6 - 7 | 1549 | 91 |