The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106311.1 | NP_001099781.1 | 6 - 7 | 1303 | 133 |
XM_006230524.2 | XP_006230586.2 | 5 - 6 | 1278 | 133 |