The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025679.1 | NP_001020850.1 | 2 - 3 | 605 | 133 |
XM_006228417.1 | XP_006228479.1 | 1 - 2 | 347 | 133 | |
GenBank mRNA | BC097499.1 | - | 2 - 3 | 605 | 133 |