Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp82
Entrez Gene ID
100910231
Gene Name
zinc finger protein 82
Gene Aliases
-
Chromosome Location
Chr.19: 14453498 - 14474286 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006255148.2 XP_006255210.1 6 - 7 897 108
XM_006255151.2 XP_006255213.1 7 - 8 1101 108

Gene Symbol
LOC102547241
Entrez Gene ID
102547241
Gene Name
zinc finger protein 82-like
Gene Aliases
-
Chromosome Location
Chr.1: 88372676 - 88400032 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006228757.2 XP_006228819.1 6 - 7 897 108
XM_006228758.2 XP_006228820.1 7 - 8 1105 108
XM_006228759.2 XP_006228821.1 7 - 8 1101 108
XM_008759232.1 XP_008757454.1 6 - 7 616 108

View Full Product Details