Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samsn1
Entrez Gene ID
170637
Gene Name
SAM domain, SH3 domain and nuclear localization signals, 1
Gene Aliases
Nash
Chromosome Location
Chr.11: 14254047 - 14304603 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130821.1 NP_570834.1 7 - 8 918 63
GenBank mRNA AB067679.1 - 7 - 8 918 63

View Full Product Details