Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thumpd2
Entrez Gene ID
313851
Gene Name
THUMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 3743080 - 3794505 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012108.1 NP_001012108.2 6 - 7 952 107
NM_001163506.1 NP_001156978.1 8 - 9 1093 107
XM_006239672.2 XP_006239734.1 8 - 9 1097 107
XM_006239673.2 XP_006239735.1 6 - 7 956 107
XM_006239675.2 XP_006239737.1 8 - 9 1097 107
XM_006239677.2 XP_006239739.1 6 - 7 956 107
XM_008764440.1 XP_008762662.1 8 - 9 1097 107
GenBank mRNA BC089870.1 - 6 - 7 952 107

View Full Product Details