Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Coro1c
Entrez Gene ID
501841
Gene Name
coronin, actin binding protein 1C
Gene Aliases
RGD1564490
Chromosome Location
Chr.12: 48481750 - 48554052 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109327.1 NP_001102797.1 10 - 11 1431 106

View Full Product Details