Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hnf1a
Entrez Gene ID
24817
Gene Name
HNF1 homeobox A
Gene Aliases
HNF1, LF-B1, Lfb1, Tcf1
Chromosome Location
Chr.12: 47407811 - 47433342 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012669.1 NP_036801.1 9 - 10 1911 109
GenBank mRNA J03170.1 - 9 - 10 1911 109
X53297.1 - 8 - 9 1269 109
X54423.1 - 9 - 10 1761 109

View Full Product Details