Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sap25
Entrez Gene ID
288559
Gene Name
Sin3A-associated protein 25
Gene Aliases
RGD1562406
Chromosome Location
Chr.12: 22125568 - 22126671 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769160.1 XP_008767382.1 1 - 2 179 138

View Full Product Details