Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crybb1
Entrez Gene ID
25421
Gene Name
crystallin, beta B1
Gene Aliases
CRYB1, CRYB11
Chromosome Location
Chr.12: 50390939 - 50404550 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012936.2 NP_037068.2 1 - 2 133 54
GenBank mRNA AW919139.1 - 1 - 2 133 54

View Full Product Details