Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tas2r108
Entrez Gene ID
554302
Gene Name
taste receptor, type 2, member 108
Gene Aliases
T2R16, Tas2r4
Chromosome Location
Chr.4: 68656928 - 68657842 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024686.1 NP_001019857.1 1 - 1 818 96
GenBank mRNA AY916510.1 - 1 - 1 818 96

View Full Product Details