Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Irf5
Entrez Gene ID
296953
Gene Name
interferon regulatory factor 5
Gene Aliases
-
Chromosome Location
Chr.4: 56804318 - 56816271 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106586.1 NP_001100056.1 7 - 8 1217 109
XM_006236180.2 XP_006236242.1 6 - 7 2370 109
XM_006236183.2 XP_006236245.1 7 - 8 1513 109
XM_006236184.2 XP_006236246.1 7 - 8 1253 109
XM_006236185.2 XP_006236247.1 7 - 8 2304 109
XM_008762765.1 XP_008760987.1 7 - 8 1170 109
GenBank mRNA FQ227572.1 - 7 - 8 1209 109
FQ230949.1 - 2 - 3 438 109

View Full Product Details