Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tcrb
Entrez Gene ID
24820
Gene Name
T-cell receptor beta chain
Gene Aliases
RATTCB, RATTCBC1, TCB, TCBC1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC105831.1 - - 100 96
BC158832.1 - - 98 96

View Full Product Details