Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpk1
Entrez Gene ID
680668
Gene Name
thiamin pyrophosphokinase 1
Gene Aliases
-
Chromosome Location
Chr.4: 72792252 - 73174179 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134994.1 NP_001128466.1 7 - 8 663 105
XM_008762921.1 XP_008761143.1 4 - 5 2209 105
GenBank mRNA BC168695.1 - 7 - 8 552 105

View Full Product Details