The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053789.1 | NP_446241.1 | 2 - 3 | 357 | 112 |
GenBank mRNA | AF218724.1 | - | 1 - 2 | 127 | 112 |