Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Seh1l
- Entrez Gene ID
-
307347
- Gene Name
- SEH1-like (S. cerevisiae)
- Gene Aliases
-
Da1-6
- Chromosome Location
-
Chr.18: 63572088 - 63656302 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY325252.1 |
- |
- |
2055 |
69 |
- Gene Symbol
- LOC100911798
- Entrez Gene ID
-
100911798
- Gene Name
- nucleoporin SEH1-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 63545029 - 63568101 on Build Rnor_6.0
View Full Product Details