Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Coro1b
Entrez Gene ID
29474
Gene Name
coronin, actin-binding protein, 1B
Gene Aliases
-
Chromosome Location
Chr.1: 219397827 - 219403253 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019222.1 NP_062095.1 7 - 8 880 153
XM_006230773.2 XP_006230835.1 7 - 8 999 153
XM_006230774.2 XP_006230836.1 7 - 8 964 153
GenBank mRNA AJ006064.1 - 7 - 8 880 153
BC061558.1 - 6 - 7 1478 153
FQ213235.1 - 3 - 4 253 153

View Full Product Details